Learn More
Medchemexpress LLC VH 101, thiol | 2358775-81-0 | 98.7% | C25H31FN4O4S2 | 25 MG
SDP
Catalog No. 5000358989
Encompass_Preferred
VH 101, thiol (compound 20) is a VHL ligand primarily used for the synthesis of PROTACs, targeting VHL. This product is intended for research use only and has not been fully validated for medical applications.
  • VHL ligand for PROTAC synthesis
  • Targets VHL
  • For research use only
Catalog No. 50-003-58989 Supplier Medchemexpress LLC Supplier No. 25MGHY47851
VH 101, thiol (compound 20) is a VHL ligand primarily used for the synthesis of PROTACs, targeting VHL. This product is intended for research use only and has not been fully validated for medical applications.
  • VHL ligand for PROTAC synthesis
  • Targets VHL
  • For research use only