Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VH 101, thiol | 2358775-81-0 | 98.7% | C25H31FN4O4S2 | 50 MG
SDP

Catalog No. 5000357891
Encompass_Preferred

VH 101, thiol (compound 20) is a VHL ligand primarily used for the synthesis of PROTACs. This product is intended for research purposes only and not for patient use.

  • VHL ligand for PROTAC synthesis
  • Intended for research use only
  • Molecular weight: 534.67
  • Appearance: solid
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 100 mg/mL

Catalog No. 50-003-57891 Supplier Medchemexpress LLC Supplier No. HY4785150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

VH 101, thiol (compound 20) is a VHL ligand primarily used for the synthesis of PROTACs. This product is intended for research purposes only and not for patient use.

  • VHL ligand for PROTAC synthesis
  • Intended for research use only
  • Molecular weight: 534.67
  • Appearance: solid
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.