Learn More
Vector Laboratories Human VH L / Von Hippel Lindau (Sandwich ELISA) ELISA Kit - LS-F12950
SDP
Supplier:  Vector Laboratories LSF129501
Encompass
LS-F12950 is a 96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Human VHL / Von Hippel Lindau in samples of Plasma and Serum. It is based upon a Sandwich assay principle and can be used to detect levels of VHL / Von Hippel Lindau as low as 0.156 nanograms per millilter.
Catalog No. NC2937953