Learn More
TARGETMOL CHEMICALS INC VH032 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2910610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. VH032 commonly used as a precursor to a PROTAC that hijacks VHL as the E3 ubiquitin ligase component.VH032 is a VHL/HIF-1(alpha) interaction inhibitor with a KdPROTACs which can recruit von Hippel-Lindau (VHL) proteins as a ligand of VHL. purity: 99%
Catalog No. 50-258-0464