Learn More
Medchemexpress LLC VH032-cyclopropane-F | 2306193-99-5 | 99.92% | 532.63 | 250 MG
SDP
Catalog No. 5000439319
Encompass_Preferred
VH032-cyclopropane-F is a VHL ligand based on VH032. It can be linked to other protein ligands (e.g., SMARCA BD ligand) via a linker to create PROTACs (e.g., PROTAC 1). PROTAC 1 functions as a partial degrader of SMARCA2 and SMARCA4.
  • Is a VH032-based VHL ligand.
  • Can be connected to other protein ligands to form PROTACs.
  • Increases FAK expression levels in A427 cells at 5 μM for 24 hours.
  • Available in solid and solution forms.
  • High purity of 99.92%.
Catalog No. 50-004-39319 Supplier Medchemexpress LLC Supplier No. 250MGHY125905
VH032-cyclopropane-F is a VHL ligand based on VH032. It can be linked to other protein ligands (e.g., SMARCA BD ligand) via a linker to create PROTACs (e.g., PROTAC 1). PROTAC 1 functions as a partial degrader of SMARCA2 and SMARCA4.
  • Is a VH032-based VHL ligand.
  • Can be connected to other protein ligands to form PROTACs.
  • Increases FAK expression levels in A427 cells at 5 μM for 24 hours.
  • Available in solid and solution forms.
  • High purity of 99.92%.