Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vh032-cyclopropane-F | 2306193-99-5 | MFCD32201089 | ≥95.0% | 532.63 | C26H33FN4O5S | 10MG
SDP

Catalog No. 5000235930
Encompass_Preferred

VH032-cyclopropane-F is a VH032-based von Hippel-Lindau (VHL) ligand used as an E3 ligase recruiter in PROTAC design and synthesis. The compound contains a functionalized cyclopropane terminus that facilitates conjugation to target ligands via linkers, enabling construction of bifunctional degraders and in vitro evaluation of VHL-mediated activity.

  • Used as an E3 ligase (VHL) ligand for PROTAC design and synthesis.
  • Molecular formula C26H33FN4O5S; molecular weight 532.63.
  • CAS number 2306193-99-5 for compound identification.
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic) and formulatable for in vivo at ≥2.5 mg/mL in common vehicle systems.
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • Purity typically ≥95% (supplier dependent).
  • Supplied in small research quantities such as 10 mg vials for synthetic applications.

Catalog No. 50-002-35930 Supplier Medchemexpress LLC Supplier No. HY12590510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VH032-cyclopropane-F is a VH032-based von Hippel-Lindau (VHL) ligand used as an E3 ligase recruiter in PROTAC design and synthesis. The compound contains a functionalized cyclopropane terminus that facilitates conjugation to target ligands via linkers, enabling construction of bifunctional degraders and in vitro evaluation of VHL-mediated activity.

  • Used as an E3 ligase (VHL) ligand for PROTAC design and synthesis.
  • Molecular formula C26H33FN4O5S; molecular weight 532.63.
  • CAS number 2306193-99-5 for compound identification.
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic) and formulatable for in vivo at ≥2.5 mg/mL in common vehicle systems.
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • Purity typically ≥95% (supplier dependent).
  • Supplied in small research quantities such as 10 mg vials for synthetic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.