Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VH032-cyclopropane-F | 2306193-99-5 | MFCD32201089 | >95.0% | 532.63 | C26H33FN4O5S | 10MM 1ML
SDP

Catalog No. 5000235996
Encompass_Preferred

VH032-cyclopropane-F is a VHL E3 ligase ligand used as a building block for PROTAC design and for ligand-E3 ligase binding studies. It is supplied as a 10 mM solution in DMSO (1 mL) and as a solid powder, enabling direct use in assay development or conjugation chemistry.

  • Provided as 10 mM solution in DMSO (1 mL) and as powder.
  • Molecular weight 532.63.
  • Chemical formula C26H33FN4O5S.
  • Purity ≥95%.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for use in PROTAC construction and E3 ligase binding studies.

Catalog No. 50-002-35996 Supplier Medchemexpress LLC Supplier No. HY12590510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VH032-cyclopropane-F is a VHL E3 ligase ligand used as a building block for PROTAC design and for ligand-E3 ligase binding studies. It is supplied as a 10 mM solution in DMSO (1 mL) and as a solid powder, enabling direct use in assay development or conjugation chemistry.

  • Provided as 10 mM solution in DMSO (1 mL) and as powder.
  • Molecular weight 532.63.
  • Chemical formula C26H33FN4O5S.
  • Purity ≥95%.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for use in PROTAC construction and E3 ligase binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.