Learn More
Medchemexpress LLC Vh032-cyclopropane-f | 2306193-99-5 | MFCD32201089 | ≥95.0% | 532.63 | C26H33FN4O5S | 50MG
SDP
Catalog No. 5000237976
Encompass_Preferred
VH032-cyclopropane-F is a von Hippel-Lindau (VHL) E3 ligase ligand used as a building block for PROTACs and targeted protein degradation. It features a hydroxyl-functionalized site for linker attachment and is formulated for both in vitro and in vivo studies.
  • VHL E3 ligase ligand suitable for PROTAC design and conjugation.
  • Hydroxyl-functionalized site for linker attachment.
  • Soluble in DMSO at 100 mg/mL (187.75 mM; may require sonication) for in vitro use.
  • Formulatable for in vivo dosing at ≥2.5 mg/mL in common vehicles.
  • Powder stable at -20°C for long-term storage; solutions stable at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-37976 Supplier Medchemexpress LLC Supplier No. HY12590550MG
VH032-cyclopropane-F is a von Hippel-Lindau (VHL) E3 ligase ligand used as a building block for PROTACs and targeted protein degradation. It features a hydroxyl-functionalized site for linker attachment and is formulated for both in vitro and in vivo studies.
  • VHL E3 ligase ligand suitable for PROTAC design and conjugation.
  • Hydroxyl-functionalized site for linker attachment.
  • Soluble in DMSO at 100 mg/mL (187.75 mM; may require sonication) for in vitro use.
  • Formulatable for in vivo dosing at ≥2.5 mg/mL in common vehicles.
  • Powder stable at -20°C for long-term storage; solutions stable at -80°C (6 months) or -20°C (1 month).