Learn More
Medchemexpress LLC VH032-cyclopropane-F | 2306193-99-5 | MFCD32201089 | 99.9% | 532.63 | C26H33FN4O5S | 5MG
SDP
Catalog No. 5000235566
Encompass_Preferred
VH032-cyclopropane-F is a VH032-based von Hippel-Lindau (VHL) E3 ligase ligand used as the VHL-recruiting moiety in targeted protein degradation research (PROTACs). It is supplied as a solid research reagent with characterized physical and chemical properties and is designed for use in synthesis and biochemical assay development.
  • High purity reported (99.92%) for research applications.
  • Available in multiple small-scale pack sizes for synthesis and testing.
  • Stable as a powder at -20°C and in solvent under recommended storage conditions.
  • Molecular weight 532.63 and formula C26H33FN4O5S for unambiguous identification.
  • Intended for use as a VHL-recruiting building block in degrader design.
Catalog No. 50-002-35566 Supplier Medchemexpress LLC Supplier No. HY1259055MG
VH032-cyclopropane-F is a VH032-based von Hippel-Lindau (VHL) E3 ligase ligand used as the VHL-recruiting moiety in targeted protein degradation research (PROTACs). It is supplied as a solid research reagent with characterized physical and chemical properties and is designed for use in synthesis and biochemical assay development.
  • High purity reported (99.92%) for research applications.
  • Available in multiple small-scale pack sizes for synthesis and testing.
  • Stable as a powder at -20°C and in solvent under recommended storage conditions.
  • Molecular weight 532.63 and formula C26H33FN4O5S for unambiguous identification.
  • Intended for use as a VHL-recruiting building block in degrader design.