Learn More
Medchemexpress LLC Vh032-cyclopropane-f | 2306193-99-5 | MFCD32201089 | >95.0% | 532.63 g/mol | C26H33FN4O5S | 100MG
SDP
Catalog No. 5000239566
Encompass_Preferred
VH032-cyclopropane-F is a VH032-based ligand for the von Hippel-Lindau (VHL) E3 ubiquitin ligase used as a building block in PROTAC and E3 ligase research. The compound has molecular formula C26H33FN4O5S, molecular weight 532.63 g/mol, and CAS 2306193-99-5. It is provided as a research-grade small molecule; handle according to applicable safety data and laboratory practices.
  • VHL E3 ligase ligand suitable for PROTAC assembly.
  • Molecular formula C26H33FN4O5S and Mw 532.63 g/mol.
  • High purity (≥95.0%) appropriate for biochemical studies.
  • Solid material for dissolution in common organic solvents.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-39566 Supplier Medchemexpress LLC Supplier No. HY125905100MG
VH032-cyclopropane-F is a VH032-based ligand for the von Hippel-Lindau (VHL) E3 ubiquitin ligase used as a building block in PROTAC and E3 ligase research. The compound has molecular formula C26H33FN4O5S, molecular weight 532.63 g/mol, and CAS 2306193-99-5. It is provided as a research-grade small molecule; handle according to applicable safety data and laboratory practices.
  • VHL E3 ligase ligand suitable for PROTAC assembly.
  • Molecular formula C26H33FN4O5S and Mw 532.63 g/mol.
  • High purity (≥95.0%) appropriate for biochemical studies.
  • Solid material for dissolution in common organic solvents.
  • Intended for research use only; not for human or veterinary use.