Learn More
Medchemexpress LLC VH032-PEG3-acetylene | 2098799-80-3 | 98.1% | 25 MG
SDP
Catalog No. 5000343038
Encompass_Preferred
VH032-PEG3-acetylene is a synthesized E3 ligase ligand-linker conjugate that incorporates the VH032 based VHL ligand and a linker used in PROTAC technology. This compound is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a VH032 based VHL ligand
  • Utilizes a linker employed in PROTAC technology
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-43038 Supplier Medchemexpress LLC Supplier No. 25MGHY128767
VH032-PEG3-acetylene is a synthesized E3 ligase ligand-linker conjugate that incorporates the VH032 based VHL ligand and a linker used in PROTAC technology. This compound is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a VH032 based VHL ligand
  • Utilizes a linker employed in PROTAC technology
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups