Learn More
Medchemexpress LLC HY-128767 5mg Medchemexpress, VH032-PEG3-acetylene CAS:2098799-80-3 Purity:>98%
SDP
Catalog No. 501969770
Encompass_Preferred
Medchemexpress, HY-128767 5mg VH032-PEG3-acetylene CAS:2098799-80-3 VH032-PEG3-acetylene is a synthesized E3 ligase ligand-linker conjugate that incorporates the VH032 based VHL ligand and a linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9770 Supplier Medchemexpress LLC Supplier No. HY1287675MG
Medchemexpress, HY-128767 5mg VH032-PEG3-acetylene CAS:2098799-80-3 VH032-PEG3-acetylene is a synthesized E3 ligase ligand-linker conjugate that incorporates the VH032 based VHL ligand and a linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.