Learn More
Medchemexpress LLC HY-112495 10mg , VH032-PEG5-C6-Cl CAS:1799506-06-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11249510MG
Encompass_Preferred
HY-112495 10mg VH032-PEG5-C6-Cl CAS: 1799506-06-1 VH032-PEG5-C6-Cl (HaloPROTAC 2) is a conjugate of ligands for E3 and 21-atom-length linker. The connector of linker is Halogen group. VH032-PEG5-C6-Cl incorporates the VH032 based VHL ligand and 5-unit PEG linker. VH032-PEG5-C6-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9943