Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VH032-PEG5-C6-Cl | 1799506-06-1 | 99.9% | 783.41 | 25 MG
SDP

Catalog No. 5000439540
Encompass_Preferred

VH032-PEG5-C6-Cl (HaloPROTAC 2) is a conjugate of ligands for E3 and a 21-atom-length linker. It incorporates a VH032-based VHL ligand and a 5-unit PEG linker, capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.

  • Target: VHL
  • Activity: Induces nearly 70% degradation of GFP-Halotag7 at 2.5 μM after 24 hours of treatment in in vitro assays
  • Application: For research use only

Catalog No. 50-004-39540 Supplier Medchemexpress LLC Supplier No. 25MGHY112495
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VH032-PEG5-C6-Cl (HaloPROTAC 2) is a conjugate of ligands for E3 and a 21-atom-length linker. It incorporates a VH032-based VHL ligand and a 5-unit PEG linker, capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.

  • Target: VHL
  • Activity: Induces nearly 70% degradation of GFP-Halotag7 at 2.5 μM after 24 hours of treatment in in vitro assays
  • Application: For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.