Learn More
Medchemexpress LLC VH032-PEG5-C6-Cl (HaloPROTAC 2) | 1799506-06-1 | 99.9% | 783.41 | 50 MG
SDP
Catalog No. 5000436331
Encompass_Preferred
VH032-PEG5-C6-Cl, also known as HaloPROTAC 2, is a conjugate of E3 ligase ligands and a 21-atom-length linker. This compound incorporates the VH032 based VHL ligand and a 5-unit PEG linker.
  • Conjugate of ligands for E3.
  • Features a 21-atom-length linker with a halogen group connector.
  • Incorporates a VH032 based VHL ligand.
  • Includes a 5-unit PEG linker.
  • Capable of inducing degradation of GFP-HaloTag7 in cell-based assays.
Catalog No. 50-004-36331 Supplier Medchemexpress LLC Supplier No. HY11249550MG
VH032-PEG5-C6-Cl, also known as HaloPROTAC 2, is a conjugate of E3 ligase ligands and a 21-atom-length linker. This compound incorporates the VH032 based VHL ligand and a 5-unit PEG linker.
  • Conjugate of ligands for E3.
  • Features a 21-atom-length linker with a halogen group connector.
  • Incorporates a VH032 based VHL ligand.
  • Includes a 5-unit PEG linker.
  • Capable of inducing degradation of GFP-HaloTag7 in cell-based assays.