Learn More
Medchemexpress LLC VH032 thiol | 2098836-54-3 | 490.64 | 1 MG
SDP
Catalog No. 5000440002
Encompass_Preferred
VH032 thiol (VHL ligand 6) is a VHL ligand that binds to pan-BET inhibitor JQ1 via a linker to form PROTAC. The VHL protein is a substrate recognition subunit of two ubiquitously expressed and biologically important Cullin RING E3 ubiquitin ligase complexes. VHL is one of the most popular E3 ligases being recruited by bifunctional Proteolysis-targeting chimeras (PROTACs) to induce ubiquitination and subsequent proteasomal degradation of a target protein.
  • Binds to pan-BET inhibitor JQ1 via a linker to form PROTAC.
  • Recruits VHL protein, a substrate recognition subunit of Cullin RING E3 ubiquitin ligase complexes.
  • Induces ubiquitination and subsequent proteasomal degradation of target proteins.
Catalog No. 50-004-40002 Supplier Medchemexpress LLC Supplier No. HY1118231MG
VH032 thiol (VHL ligand 6) is a VHL ligand that binds to pan-BET inhibitor JQ1 via a linker to form PROTAC. The VHL protein is a substrate recognition subunit of two ubiquitously expressed and biologically important Cullin RING E3 ubiquitin ligase complexes. VHL is one of the most popular E3 ligases being recruited by bifunctional Proteolysis-targeting chimeras (PROTACs) to induce ubiquitination and subsequent proteasomal degradation of a target protein.
  • Binds to pan-BET inhibitor JQ1 via a linker to form PROTAC.
  • Recruits VHL protein, a substrate recognition subunit of Cullin RING E3 ubiquitin ligase complexes.
  • Induces ubiquitination and subsequent proteasomal degradation of target proteins.