Learn More
Medchemexpress LLC VH032 thiol | 2098836-54-3 | >95.0% | 490.64 | 10 MG
SDP
Catalog No. 5000438956
Encompass_Preferred
VH032 thiol (VHL ligand 6) is a VHL ligand designed to bind to the pan-BET inhibitor JQ1 via a linker, forming a PROTAC. This product is intended for research use only and is not sold to patients.
  • Target: Ligands for E3 Ligase
  • Pathway: PROTAC
  • IC50 & target: VHL
Catalog No. 50-004-38956 Supplier Medchemexpress LLC Supplier No. HY11182310MG
VH032 thiol (VHL ligand 6) is a VHL ligand designed to bind to the pan-BET inhibitor JQ1 via a linker, forming a PROTAC. This product is intended for research use only and is not sold to patients.
  • Target: Ligands for E3 Ligase
  • Pathway: PROTAC
  • IC50 & target: VHL