Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VH032 thiol (VHL ligand 6) | 2098836-54-3 | 95.9% | 490.64 g/mol | C23H30N4O4S2 | 5 MG
SDP

Catalog No. 5000264146
Encompass_Preferred

VH032 thiol (VHL ligand 6) is a von Hippel-Lindau (VHL) E3 ligase ligand used as an E3-recruiting moiety in PROTAC design and degrader development. It is characterized by a molecular formula of C23H30N4O4S2 and a molecular weight of 490.64 g/mol. Provided for research use only, the material is supplied as a solid with documented solubility and storage recommendations.

  • Used as an E3 ligase recruiting moiety in PROTAC synthesis.
  • Facilitates conjugation to target ligands via thiol-reactive linkers.
  • Supplied as a white to light yellow solid with ~95% purity.
  • Soluble in DMSO at 50 mg/mL; example in vivo formulations are provided.
  • Recommended storage at 4°C under inert atmosphere; in-solution storage conditions documented.

Catalog No. 50-002-64146 Supplier Medchemexpress LLC Supplier No. HY1118235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VH032 thiol (VHL ligand 6) is a von Hippel-Lindau (VHL) E3 ligase ligand used as an E3-recruiting moiety in PROTAC design and degrader development. It is characterized by a molecular formula of C23H30N4O4S2 and a molecular weight of 490.64 g/mol. Provided for research use only, the material is supplied as a solid with documented solubility and storage recommendations.

  • Used as an E3 ligase recruiting moiety in PROTAC synthesis.
  • Facilitates conjugation to target ligands via thiol-reactive linkers.
  • Supplied as a white to light yellow solid with ~95% purity.
  • Soluble in DMSO at 50 mg/mL; example in vivo formulations are provided.
  • Recommended storage at 4°C under inert atmosphere; in-solution storage conditions documented.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.