Learn More
Selleck Chemical LLC VH298-5mg
SDP
Catalog No. 502389855
Encompass_Preferred
VH298 is a potent VHL (Von Hippel-Lindau, the E3 ligase) inhibitor that stabilizes HIF-α. VH298 blocks the VHL:HIF-α interaction with Kd of 90 nM in isothermal titration calorimetry (ITC) and 80 nM in a competitive fluorescence polarization assay. VH-298 can be used in PROTAC technology.
Catalog No. 50-238-9855 Supplier Selleck Chemical LLC Supplier No. S84495MG
VH298 is a potent VHL (Von Hippel-Lindau, the E3 ligase) inhibitor that stabilizes HIF-α. VH298 blocks the VHL:HIF-α interaction with Kd of 90 nM in isothermal titration calorimetry (ITC) and 80 nM in a competitive fluorescence polarization assay. VH-298 can be used in PROTAC technology.