Learn More
Amsco Scientific Steris VH2O2 Indicator Tape
Catalog No. NC3316273
Encompass
VH2O2 Indicator Tape Six rolls/case. The VERIFY VH2O2 Indicator Tape provides steadfast adhesion of synthetic wrap for hydrogen peroxide sterilization. The indicator ink is designed to be easy-to-read and provides immediate identification of processed packs.
This item is not returnable. View return policy
Catalog No. NC3316273 Supplier Amsco Scientific Steris Supplier No. PCC071
VH2O2 Indicator Tape Six rolls/case. The VERIFY VH2O2 Indicator Tape provides steadfast adhesion of synthetic wrap for hydrogen peroxide sterilization. The indicator ink is designed to be easy-to-read and provides immediate identification of processed packs.