Learn More
Medchemexpress LLC Vh4127 TFA | 1034.35 (free base) | 1 MG
SDP
Catalog No. 5000425552
Encompass_Preferred
VH4127 TFA is a cyclic peptide that targets the low-density lipoprotein receptor (LDLR) with a KD of 18 nM for hLDLR. It specifically binds to the epidermal growth factor (EGF) homology domain of both rodent and human LDLR.
  • Targets low density lipoprotein receptor (LDLR)
  • Has a high binding affinity (KD of 18 nM) for human LDLR
  • Binds specifically to the EGF homology domain of rodent and human LDLR
  • Available in various quantities
  • For research use only
  • Information on solubility, in vivo dissolution, and related antibodies is also available
Catalog No. 50-004-25552 Supplier Medchemexpress LLC Supplier No. HYP10697A1MG
VH4127 TFA is a cyclic peptide that targets the low-density lipoprotein receptor (LDLR) with a KD of 18 nM for hLDLR. It specifically binds to the epidermal growth factor (EGF) homology domain of both rodent and human LDLR.
  • Targets low density lipoprotein receptor (LDLR)
  • Has a high binding affinity (KD of 18 nM) for human LDLR
  • Binds specifically to the EGF homology domain of rodent and human LDLR
  • Available in various quantities
  • For research use only
  • Information on solubility, in vivo dissolution, and related antibodies is also available