Learn More
3M™ VHB™ Heavy Duty Mounting Tape
Catalog No. 19072086 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
15 yd. x 3/4 in.
Catalog No. Dimensions (L x W)
19-072-086 15 yd. x 3/4 in.
1 options
Catalog No. 19-072-086 Supplier 3M™ Supplier No. 7100047923
Add to Cart
Add to Cart
3M™ VHB™ Heavy Duty Mounting Tape is a general purpose acrylic adhesive on both sides of a firm, foam core
  • Fast and easy-to-use permanent bonding method provides high strength and long-term durability
  • Offers design flexibility with its viscoelasticity and powerful ability to bond to a variety of surfaces.
  • Virtually invisible fastening keeps surfaces smooth
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives for transparent applications
  • Clear, 0.040 in (1.0 mm), general purpose adhesive and firm acrylic foam core offers a good balance of strength and conformability
  • Eliminate drilling, grinding, refinishing, screwing, welding and associated clean-up
  • Creates a permanent seal against water, moisture and more
  • Pressure sensitive adhesive bonds on contact to provide immediate handling strength
  • Allows the use of thinner, lighter weight and dissimilar materials
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mounting Tape
For Use With (Application) Commericial Solutions, Communications, Design and Construction, Electronics, Energy, Manufacturing, Safety, Transportation
Color Clear
Dimensions (L x W) 15 yd. x 3/4 in.
Model 4910
Quantity 12 Rolls/Cs.
Thickness (English) 40 mil