Learn More
3M™ VHB™ Heavy Duty Mounting Tape
Catalog No. 19072032 Shop All 3M Products
Change view
Click to view available options
Quantity:
12 Rolls/Cs.
Catalog No. Quantity
19-072-032 12 Rolls/Cs.
1 options
Catalog No. 19-072-032 Supplier 3M™ Supplier No. 7000049237
Add to Cart
Add to Cart
3M™ VHB™ Heavy Duty Mounting Tape is a fast and easy-to-use permanent bonding method that provides high strength and long-term durability.
  • Offers design flexibility with its viscoelasticity and powerful ability to bond a variety of surfaces.
  • Virtually invisible fastening keeps surfaces smooth
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives
  • 0.045 in (1.1 mm), modified acrylic adhesive
  • 45 mil thick
  • 3/4 in. x 15 yd.
  • Conformable acrylic foam core bonds to a wide variety of substrates including powder coated paints and irregular surfaces
  • Eliminate drilling, grinding, refinishing, screwing, welding and clean-up
  • Creates a permanent seal against water, moisture and more by offering better gap filling capabilities
  • Pressure sensitive adhesive bonds on contact to provide immediate handling strength
  • Allows the use of thinner, lighter weight and dissimilar materials
TRUSTED_SUSTAINABILITY
Specifications
Product Type Heavy Duty Mounting Tape
For Use With (Application) Bonding Glass to a Metal Clothes Washer Door Stand Decorative Trim Attachment, Bonding Panels to Metal Frames, Commercial Vehicle Roof Vent Attachment, Gasket Assembly, Interior Rail Car Construction, Mounting and Trim Attachment, Panel to Frame/Stiffner to Panel Assembly, TV
Color Black
Model 5952
Quantity 12 Rolls/Cs.