Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ VHB™ Tape

Catalog No. p-7182401 Shop All 3M Products
Change view
Click to view available options
Color:
Dark Gray
Gray
Adhesive Type:
Modified Acrylic
Multi-purpose Acrylic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Adhesive Type
19-072-039 Dark Gray Modified Acrylic
19-072-087 Gray Multi-purpose Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-039 Supplier 3M™ Supplier No. 7000123924
Only null left
Add to Cart
Add to Cart

3M™ VHB™ Tape is a fast and easy to use permanent bonding method that provides high strength and long-term durability. It offers design flexibility with its viscoelasticity and powerful ability to bond to a variety of surfaces.

  • Fast and easy-to-use permanent bonding method provides high strength and long-term durability
  • Virtually invisible fastening keeps surfaces smooth
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
  • Eliminate drilling, grinding, refinishing, screwing, welding and associated clean-up
  • Creates a permanent seal against water, moisture and more
  • Pressure sensitive adhesive bonds on contact to provide immediate handling strength
  • Allows the use of thinner, lighter weight and dissimilar materials
TRUSTED_SUSTAINABILITY

Specifications

Product Type VHB Tape
For Use With (Application) Bonding Panels to Metal Frames
Adhesive Type Modified Acrylic
Color Dark Gray
Dimensions (L x W) 36 yd. x 1 in.
Material Acrylic Foam Core
Model 4611
Quantity 9 Rolls/Cs.
Thickness (English) 45 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.