Learn More
Mitegen Replacement Canister For The VHC35 or TW35 IC Biomedical Storage Dewars
SDP
Catalog No. NC2259468
Encompass
This replacement canister is specifically designed for seamless compatibility with the IC Biomedical VHC35 Storage Refrigerators and TW35 Freezers, ensuring continued optimal performance for your cryogenic sample management. Constructed from durable, thin-gauge stainless steel, this 11-inch canister provides a reliable and robust solution for organizing and securely holding your valuable samples within cryogenic storage units. It plays a crucial role in maintaining the cryogenic environment by supporting efficient liquid nitrogen distribution and ensuring stable temperatures for long-term sample viability.
Catalog No. NC2259468 Supplier Mitegen Supplier No. TWR0369C24
This replacement canister is specifically designed for seamless compatibility with the IC Biomedical VHC35 Storage Refrigerators and TW35 Freezers, ensuring continued optimal performance for your cryogenic sample management. Constructed from durable, thin-gauge stainless steel, this 11-inch canister provides a reliable and robust solution for organizing and securely holding your valuable samples within cryogenic storage units. It plays a crucial role in maintaining the cryogenic environment by supporting efficient liquid nitrogen distribution and ensuring stable temperatures for long-term sample viability.