Learn More
Cayman Chemical VHL Ligand 1 (hydrochloride), Purity: ≥95%, CAS: 1448189-80-7, 100 mg
SDP
Catalog No. 502986386
Encompass_Preferred
A PROTAC building block; VHL E3 binding portion of BET PROTAC MZ1
Catalog No. 50-298-6386 Supplier Cayman Chemical Supplier No. 21591100 MG
A PROTAC building block; VHL E3 binding portion of BET PROTAC MZ1