Learn More
Cayman Chemical VHL LIgnd 1hydrochlorIde 25mg
SDP
Catalog No. 502964259
Encompass_Preferred
A PROTAC building block; VHL E3 binding portion of BET PROTAC MZ1
Catalog No. 50-296-4259 Supplier Cayman Chemical Supplier No. 2159125 MG
A PROTAC building block; VHL E3 binding portion of BET PROTAC MZ1