Learn More
Enzo Life Sciences VHL monoclonal antibody (AT82B10) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7460100
Encompass_Preferred
Clone: AT82B10. Isotype: IgG2bκ. Immunogen: Recombinant human VHL (aa 1-154) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, ICC, IF, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3287