Learn More
IDEX Health & Science Upchurch VHP FTG 1/16IN OD LONG 10PK
Catalog No. NC1919830
Encompass_Preferred
Idex Reusable One-Piece Very High Pressure Fittings, Diameter Outer: 0.0625 in., 0.159 cm, Description: Thread: 10-32 coned; Torque: 10 in.-lbs., Stainless Steel, Proprietary PEEK Polymer Blend, VHP Fitting, Black, Pressure: 25,000 psi
Catalog No. NC1919830 Supplier IDEX Health & Science Upchurch Supplier No. VHP325X
Idex Reusable One-Piece Very High Pressure Fittings, Diameter Outer: 0.0625 in., 0.159 cm, Description: Thread: 10-32 coned; Torque: 10 in.-lbs., Stainless Steel, Proprietary PEEK Polymer Blend, VHP Fitting, Black, Pressure: 25,000 psi