Learn More
Analytical Sales & Services VHP Stainless Steel ZDV Union
Catalog No. 5000806247
Encompass
VHP Stainless Steel Union Assembly ZDV .007 thru hole
Catalog No. 50-008-06247 Supplier Analytical Sales & Services Supplier No. 64411
VHP Stainless Steel Union Assembly ZDV .007 thru hole