Learn More
Eurofins Discoverx VHR, 250 µg. (14-594M)
Supplier:  Eurofins Discoverx 14594M
Encompass
The VHR product is purified N terminal GST-tagged recombinant VHR enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 2.33 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9022