Learn More
Beckman Coulter VI-CELL BLU INSTRMT STARTUP
Catalog No. NC1920875
Encompass
  • Fully automated system for cell viability and counting
  • Eliminates manual microscopy and hemacytometer methods
  • Designed to accelerate sample processing
  • Enhances data reliability for a wide range of applications
  • Includes a comprehensive startup kit for immediate operational readiness
  • Utilizes a high-speed camera for image capture during sample flow
  • Ensures instrument-to-instrument comparability
  • Automated sample preparation and cleaning integrated
  • Data compliant with 21 CFR Part 11 regulations
  • RFID tracking for reagent management
This item is not returnable. View return policy
Catalog No. NC1920875 Supplier Beckman Coulter Supplier No. C19201
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
  • Fully automated system for cell viability and counting
  • Eliminates manual microscopy and hemacytometer methods
  • Designed to accelerate sample processing
  • Enhances data reliability for a wide range of applications
  • Includes a comprehensive startup kit for immediate operational readiness
  • Utilizes a high-speed camera for image capture during sample flow
  • Ensures instrument-to-instrument comparability
  • Automated sample preparation and cleaning integrated
  • Data compliant with 21 CFR Part 11 regulations
  • RFID tracking for reagent management