Learn More
Beckman Coulter Vi-CELL BLU Quad Pak (Qty 4)
Catalog No. NC1695436
Encompass
We’ve improved the Vi-CELL reagent pack to make it easier to use. The Vi-CELL reagents you know and trust are now packaged in a convenient ready-to-load kit. Simply open the instrument reagent panel and slide in the reagent pack. Each pack can be used for 310 activities before changing. Packs are easy to replace with no mess. Say goodbye to wrangling with tubes and splattering dye when you change out reagents. Your lab coat will thank you!Each reagent pack contains:Trypan Blue bottleWaste bottleBuffer solution bottleDisinfectant bottleCleaning agent bottle
Catalog No. NC1695436 Supplier Beckman Coulter Supplier No. C39291
We’ve improved the Vi-CELL reagent pack to make it easier to use. The Vi-CELL reagents you know and trust are now packaged in a convenient ready-to-load kit. Simply open the instrument reagent panel and slide in the reagent pack. Each pack can be used for 310 activities before changing. Packs are easy to replace with no mess. Say goodbye to wrangling with tubes and splattering dye when you change out reagents. Your lab coat will thank you!Each reagent pack contains:Trypan Blue bottleWaste bottleBuffer solution bottleDisinfectant bottleCleaning agent bottle