Learn More
CHEMOMETEC Via1-Cassette™ box with 100 pcs
SDP
Catalog No. NC1420193
Encompass
The Via1-Cassette™ is a specialized cell sampling and staining device for NucleoCounter instruments. It contains two immobilized fluorophores, acridine orange (AO) and DAPI. Approximately 60 µl of cell sample is loaded into the cassette, dissolving the AO and DAPI in the mixing channels. AO stains all cells in the sample, both live and dead cells, thus providing a total count; DAPI stains the dead cells. Each cassette carries an individual dot-code indicating the exact volume of its counting chamber. This dot-code is read by the NucleoCounter instrument at time of analysis and is used by the software to calculate cell concentration.
This item is not returnable. View return policy
Catalog No. NC1420193 Supplier CHEMOMETEC Supplier No. 9410012
The Via1-Cassette™ is a specialized cell sampling and staining device for NucleoCounter instruments. It contains two immobilized fluorophores, acridine orange (AO) and DAPI. Approximately 60 µl of cell sample is loaded into the cassette, dissolving the AO and DAPI in the mixing channels. AO stains all cells in the sample, both live and dead cells, thus providing a total count; DAPI stains the dead cells. Each cassette carries an individual dot-code indicating the exact volume of its counting chamber. This dot-code is read by the NucleoCounter instrument at time of analysis and is used by the software to calculate cell concentration.