Learn More
BIOGEMS INTERNATIONAL INC Viability Dye 780 100 tests
SDP
Catalog No. 501530218
Encompass
The Viability Dye 780 can be used to exclude non-viable cells in flow cytometry assays. It irreversibly binds to cell surface and intracellular amines. The dye can covalently bind to a higher concentration of amines in cells that have lost plasma membrane integrity, such as those in the late stages of apoptosis or dead cells, and generate a significantly more intense fluorescence signal. The advantage of this viability dye over propidium iodide and 7-AAD, is that the cells it denotes can be washed, fixed, permeabilized and intracellularly stained without losing the fluorescence of the viability staining. 
The Viability Dye 780 can be excited by the red (633nm) laser and has a peak emission of 780nm detectable by the 780/60 band pass filter.
Catalog No. 50-153-0218 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 6291000100
The Viability Dye 780 can be used to exclude non-viable cells in flow cytometry assays. It irreversibly binds to cell surface and intracellular amines. The dye can covalently bind to a higher concentration of amines in cells that have lost plasma membrane integrity, such as those in the late stages of apoptosis or dead cells, and generate a significantly more intense fluorescence signal. The advantage of this viability dye over propidium iodide and 7-AAD, is that the cells it denotes can be washed, fixed, permeabilized and intracellularly stained without losing the fluorescence of the viability staining. 
The Viability Dye 780 can be excited by the red (633nm) laser and has a peak emission of 780nm detectable by the 780/60 band pass filter.