Learn More
Cytek Biosciences Inc Viability Dyes 7-AAD ..... Cat. No. 13-6993-T200 ..... Size 200 tests
Supplier:  Cytek Biosciences Inc 136993T200
Encompass_Preferred
Viability Dyes 7-AAD ..... Cat. No. 13-6993-T200 ..... Size 200 tests7-AAD (7-Aminoactinomycin D) is a nucleic acid dye that can be used to exclude nonviable cells from the analysis of flow cytometry data.
Catalog No. 50-210-2679