Learn More
BANGS LABORATORIES INC ViaCheck™ Concentration Control, Critical Reference Material, Exceptional Stability, 20 mL
Supplier:  BANGS LABORATORIES INC VC80N
Encompass
The ViaCheck™ Concentration Control is a pre-stained microsphere suspension designed for quality control of cell viability analyzers. Mimicking live and dead cells, it ensures accurate results without sample prep, offering exceptional reproducibility and stability. Ideal for biomanufacturing, this non-biological reference material eliminates variability inherent in biological samples, enhancing instrument performance.
Catalog No. NC1279459