Learn More
Polysciences, Inc. ViaCheck Concentration Control 8 x 10^6, 20 mL
SDP
Supplier:  Polysciences, Inc. 2462920
Encompass
ViaCheck Concentration Control products are a suspension of microspheres, that are produced at a fixed concentration of beads/ml.
Catalog No. NC0252058