Learn More
Integra Biosciences Corp 16 Channel VIAFLO, Lightweight Electronic Pipette
SDP
Catalog No. NC0805687
Encompass
The VIAFLO pipette offers perfect balance, an ultra lightweight design, unsurpassed operational comfort. Easy to understand and intuitive to use, due to simple sliding with the finger over the touch wheel and full text menus.
Catalog No. NC0805687 Supplier Integra Biosciences Corp Supplier No. 4646
The VIAFLO pipette offers perfect balance, an ultra lightweight design, unsurpassed operational comfort. Easy to understand and intuitive to use, due to simple sliding with the finger over the touch wheel and full text menus.