Learn More
Biotium ViaFluor 405 Live Cell Microtubule Stain, Trial Size
Catalog No. 501964769
Encompass_Preferred
ViaFluor™ 405 is a blue fluorescent taxol probe with excitation/emission at 408/452 nm that allows fluorescent staining of microtubules in living cells for detection in the DAPI channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.
Catalog No. 50-196-4769 Supplier Biotium Supplier No. 70064
ViaFluor™ 405 is a blue fluorescent taxol probe with excitation/emission at 408/452 nm that allows fluorescent staining of microtubules in living cells for detection in the DAPI channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.