Learn More
Biotium ViaFluor 405 Live Cell Microtubule Stain
Catalog No. 501964770
Encompass_Preferred
ViaFluor™ 405 is a blue fluorescent taxol probe with excitation/emission at 408/452 nm that allows fluorescent staining of microtubules in living cells for detection in the DAPI channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.
Catalog No. 50-196-4770 Supplier Biotium Supplier No. 70064T
ViaFluor™ 405 is a blue fluorescent taxol probe with excitation/emission at 408/452 nm that allows fluorescent staining of microtubules in living cells for detection in the DAPI channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.