Learn More
Biotium ViaFluor 405 SE Cell Proliferation Kit (10 vials, 1000 assays)
Catalog No. 501964493
Encompass_Preferred
ViaFluor SE Cell Proliferation Kits use non-toxic, amine-reactive dyes to covalently label cells throughout the cell cytoplasm and intracellular compartments. Cell proliferation dyes are commonly used to monitor cell division by flow cytometry. The dyes also can be used to stably label cells to image cell morphology, or to track cell populations in mixed co-culture experiments. Labeling is covalent, so it withstands fixation and permeabilization for subsequent immunostaining. This kit contains ViaFluor 405 SE which has an excitation/emission of 408/452 nm. The kit contains 10 lyophilized dye vials. Each vial is sufficient for 20 uL of 5000X stock solution.
Catalog No. 50-196-4493 Supplier Biotium Supplier No. 30068
ViaFluor SE Cell Proliferation Kits use non-toxic, amine-reactive dyes to covalently label cells throughout the cell cytoplasm and intracellular compartments. Cell proliferation dyes are commonly used to monitor cell division by flow cytometry. The dyes also can be used to stably label cells to image cell morphology, or to track cell populations in mixed co-culture experiments. Labeling is covalent, so it withstands fixation and permeabilization for subsequent immunostaining. This kit contains ViaFluor 405 SE which has an excitation/emission of 408/452 nm. The kit contains 10 lyophilized dye vials. Each vial is sufficient for 20 uL of 5000X stock solution.