Learn More
Biotium ViaFluor 488 Live Cell Microtubule Stain
Catalog No. 501964768
Encompass_Preferred
ViaFluor™ 488 is a green fluorescent taxol probe with excitation/emission at 500/515 nm that allows fluorescent staining of microtubules in living cells for detection in the FITC channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.
Catalog No. 50-196-4768 Supplier Biotium Supplier No. 70062
ViaFluor™ 488 is a green fluorescent taxol probe with excitation/emission at 500/515 nm that allows fluorescent staining of microtubules in living cells for detection in the FITC channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.