Learn More
Biotium ViaFluor 647 Live Cell Microtubule Stain, 50uL
Catalog No. NC1669456
Encompass_Preferred
ViaFluor Live Cell Microtubule Stains are cell-permeable taxol probes for imaging the microtubule cytoskeleton in live cells. Available with blue, green, or far-red fluorescence.
Catalog No. NC1669456 Supplier Biotium Supplier No. 70063
ViaFluor Live Cell Microtubule Stains are cell-permeable taxol probes for imaging the microtubule cytoskeleton in live cells. Available with blue, green, or far-red fluorescence.