Learn More
Biotium ViaFluor CFSE Cell Proliferation Kit
Catalog No. 501964486
Encompass_Preferred
ViaFluor SE Cell Proliferation Kits use non-toxic, amine-reactive dyes to covalently label cells throughout the cell cytoplasm and intracellular compartments. Cell proliferation dyes are commonly used to monitor cell division by flow cytometry. The dyes also can be used to stably label cells to image cell morphology, or to track cell populations in mixed co-culture experiments. Labeling is covalent, so it withstands fixation and permeabilization for subsequent immunostaining. This kit contains ViaFluor CFSE which has an excitation/emission of 495/519 nm. The kit contains 10 lyophilized dye vials. Each vial is sufficient for 18 uL of 5000X stock solution.
Catalog No. 50-196-4486 Supplier Biotium Supplier No. 30050
ViaFluor SE Cell Proliferation Kits use non-toxic, amine-reactive dyes to covalently label cells throughout the cell cytoplasm and intracellular compartments. Cell proliferation dyes are commonly used to monitor cell division by flow cytometry. The dyes also can be used to stably label cells to image cell morphology, or to track cell populations in mixed co-culture experiments. Labeling is covalent, so it withstands fixation and permeabilization for subsequent immunostaining. This kit contains ViaFluor CFSE which has an excitation/emission of 495/519 nm. The kit contains 10 lyophilized dye vials. Each vial is sufficient for 18 uL of 5000X stock solution.