Learn More
Biotium ViaFluor 405 SE Cell Proliferation Kit (1 vial, 100 assays)
Catalog No. NC1540663
Encompass_Preferred
ViaFluor SE Cell Proliferation Dyes are fluorescent tracers for tracking cell division by flow cytometry; ViaFluor 405 SE Cell Proliferation Dye is an improved replacement for the classical proliferation dye CFSE. It can be detected in the Pacific Blue channel with violet laser excitation.
Catalog No. NC1540663 Supplier Biotium Supplier No. 30068T
ViaFluor SE Cell Proliferation Dyes are fluorescent tracers for tracking cell division by flow cytometry; ViaFluor 405 SE Cell Proliferation Dye is an improved replacement for the classical proliferation dye CFSE. It can be detected in the Pacific Blue channel with violet laser excitation.