Learn More
Biotium ViaFluor 488 SE Cell Proliferation Kit, Trial Size (1 vial, 100 assays)
Catalog No. NC1540664
Encompass_Preferred
ViaFluor SE Cell Proliferation Dyes are fluorescent tracers for tracking cell division by flow cytometry; ViaFluor 488 SE Cell Proliferation Dye is an improved replacement for the classical proliferation dye CFSE. It can be detected in the FITC channel with blue laser excitation.
Catalog No. NC1540664 Supplier Biotium Supplier No. 30086T
Add to Cart
Add to Cart
ViaFluor SE Cell Proliferation Dyes are fluorescent tracers for tracking cell division by flow cytometry; ViaFluor 488 SE Cell Proliferation Dye is an improved replacement for the classical proliferation dye CFSE. It can be detected in the FITC channel with blue laser excitation.