Learn More
Beckman Coulter ViaKrome 405 Fixable Viability Dye
Supplier:  Beckman Coulter C36614/DEL
Encompass
ViaKrome Fixable Viability Dyes are used for the discrimination by flow cytometry of live and dead mammalian cells based on their fluorescence intensity. Since ViaKrome Fixable Viability Dyes are covalently bound to cellular proteins, the cells can further be processed through intracellular (Fixation and Permeabilization) and/or extracellular staining procedures prior to flow cytometry analysis.
Catalog No. NC3690288