Learn More
Chemglass Life Sciences Vial, 0.1mL, TPX, Polypropylene, Limited Volume, Large Opening, 12x32mm, 9mm Thread
SDP
Catalog No. 501532430
Encompass_Preferred
Vial, 0.1mL, TPX, Polypropylene, Limited Volume, 12x32mm, 9mm Thread
Catalog No. 50-153-2430 Supplier Chemglass Life Sciences Supplier No. CV10081232
Vial, 0.1mL, TPX, Polypropylene, Limited Volume, 12x32mm, 9mm Thread