Learn More
Chemglass Life Sciences Aluminum Seal, Silver, 20mm, PTFE/Gray Butyl Rubber Septa
SDP
Catalog No. 501532536
Encompass_Preferred
Aluminum Seal, Silver, 20mm, PTFE/Gray Butyl Rubber Septa. All septa are pre-inserted to prevent contamination and for convenience.
Catalog No. 50-153-2536 Supplier Chemglass Life Sciences Supplier No. CV32000020
Aluminum Seal, Silver, 20mm, PTFE/Gray Butyl Rubber Septa. All septa are pre-inserted to prevent contamination and for convenience.